siRNA Details
| Gene Symbol: ZNF304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290318.1 | NP_001277247.1 | 4 | 3331 | ||
| NM_001290319.1 | NP_001277248.1 | 4 | 3271 | |||
| NM_020657.3 | NP_065708.2 | 3 | 3190 | |||
| XM_011527145.2 | XP_011525447.1 | 5 | 3433 | |||
| XM_017027024.1 | XP_016882513.1 | 4 | 3723 | |||
| GenBank mRNA | AJ276316.1 | - | 4 | 3085 | ||
| BC111531.1 | - | 4 | 3219 | 40012297 | ||
| BC111532.1 | - | 4 | 3279 | 40012300 | ||

ZNF304
Genomic Map