siRNA Details
| Gene Symbol: ZFP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020828.1 | NP_065879.1 | 8 | 1159 | ||
| XM_011526462.2 | XP_011524764.1 | 8 | 1155 | |||
| XM_011526463.2 | XP_011524765.2 | 8 | 1888 | |||
| GenBank mRNA | AB037852.1 | - | 8 | 1159 | ||
| AF507045.1 | - | 8 | 1159 | |||
| BC127002.1 | - | 8 | 1144 | 40108311 | ||
| BC127003.1 | - | 8 | 1144 | 40108312 | ||
| CR749659.1 | - | 437 | ||||

ZFP28
Genomic Map