siRNA Details
| Gene Symbol: HOMEZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020834.2 | NP_065885.2 | 2 | 631 | ||
| GenBank mRNA | AB037864.1 | - | 2 | 557 | ||
| AF463523.1 | - | 2 | 555 | |||
| AK296927.1 | - | 2 | 591 | |||
| AK301849.1 | - | 2 | 477 | |||
| AK302788.1 | - | 2 | 491 | |||
| BC130392.1 | - | 1 | 456 | 40146423 | ||

HOMEZ
Genomic Map