siRNA Details
| Gene Symbol: ZNF71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021216.4 | NP_067039.1 | 3 | 2484 | ||
| XM_011527193.2 | XP_011525495.1 | 4 | 2636 | |||
| XM_017027078.1 | XP_016882567.1 | 4 | 2546 | |||
| XM_017027079.1 | XP_016882568.1 | 4 | 4416 | |||
| XM_017027080.1 | XP_016882569.1 | 3 | 2419 | |||
| GenBank mRNA | BC014280.2 | - | 3 | 2456 | 3840512 | |

ZNF71
Genomic Map