siRNA Details
| Gene Symbol: SCAND2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003654.2 | 3 | 957 | |||
| NR_004859.1 | 4 | 1515 | ||||
| GenBank mRNA | AA280726.1 | - | 3 | 219 | ||
| AF229246.1 | - | 4 | 2775 | |||
| AI218957.1 | - | 3 | 219 | |||
| AK022844.1 | - | 3 | 957 | |||
| AK290489.1 | - | 3 | 967 | |||
| AL109690.1 | - | 3 | 1515 | |||
| AW341587.1 | - | 4 | 299 | |||
| BC011547.1 | - | 3 | 939 | |||
| BC012929.1 | - | 3 | 939 | |||
| BC063836.1 | - | 3 | 913 | |||

SCAND2P
Genomic Map