siRNA Details
| Gene Symbol: ZNF649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023074.3 | NP_075562.2 | 4 | 527 | ||
| GenBank mRNA | AK000909.1 | - | 4 | 463 | ||
| AK022706.1 | - | 4 | 525 | |||
| AK225480.1 | - | 4 | 486 | |||
| AK315487.1 | - | 4 | 489 | |||
| AL713677.1 | - | 4 | 469 | |||
| BC005368.1 | - | 4 | 486 | 3930849 | ||
| DA782250.1 | - | 4 | 527 | |||

ZNF649
Genomic Map