siRNA Details
| Gene Symbol: ZNF768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024671.3 | NP_078947.3 | 2 | 621 | ||
| XM_017023665.1 | XP_016879154.1 | 2 | 1626 | |||
| XM_017023666.1 | XP_016879155.1 | 2 | 446 | |||
| GenBank mRNA | AK027089.1 | - | 2 | 393 | ||
| AK295274.1 | - | 2 | 390 | |||
| AK315449.1 | - | 2 | 417 | |||
| BC013760.1 | - | 2 | 621 | 3856266 | ||
| BC092403.1 | - | 2 | 533 | 6201079 | ||

ZNF768
Genomic Map