siRNA Details
| Gene Symbol: ZNF408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184751.1 | NP_001171680.1 | 4, 5 | 647 | ||
| NM_024741.2 | NP_079017.1 | 4, 5 | 932 | |||
| GenBank mRNA | AF346626.1 | - | 4, 5 | 882 | ||
| AK022889.1 | - | 4, 5 | 881 | |||
| AK302180.1 | - | 4, 5 | 647 | |||
| BC013355.2 | - | 4, 5 | 654 | 4040144 | ||
| BC015708.2 | - | 4, 5 | 661 | 3911552 | ||

ZNF408
Genomic Map