siRNA Details
| Gene Symbol: ZNF556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300843.1 | NP_001287772.1 | 4 | 466 | ||
| NM_024967.2 | NP_079243.1 | 4 | 469 | |||
| GenBank mRNA | AK021699.1 | - | 4 | 469 | ||
| AK225297.1 | - | 4 | 466 | |||
| BC009374.2 | - | 4 | 446 | 4121795 | ||

ZNF556
Genomic Map