siRNA Details
| Gene Symbol: ZNF606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025027.3 | NP_079303.2 | 7 | 3357 | ||
| XM_005259276.1 | XP_005259333.1 | 7 | 3217 | |||
| GenBank mRNA | AB058755.1 | - | 7 | 3360 | ||
| AF455357.1 | - | 8 | 3919 | |||
| AK024322.1 | - | 1 | 1579 | |||
| BC037209.1 | - | 7 | 2831 | 4837545 | ||

ZNF606
Genomic Map