siRNA Details
| Gene Symbol: TRIM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025188.3 | NP_079464.2 | 2 | 1794 | ||
| XM_011542199.2 | XP_011540501.1 | 3 | 1090 | |||
| GenBank mRNA | AK023243.1 | - | 2 | 1733 | ||
| AK222901.1 | - | 2 | 1738 | |||
| AY669488.1 | - | 2 | 1794 | |||
| BU165629.1 | - | 1 | 522 | |||

TRIM45
Genomic Map