siRNA Details
Gene Symbol: SOX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1300 | ||
GenBank mRNA | AK055556.1 | - | 2 | 1238 | ||
AK223574.1 | - | 2 | 1238 | |||
BC004299.1 | - | 2 | 1227 | |||
BC071947.1 | - | 2 | 1226 | 6459570 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011446, BC131936, BC145925, AB023419, AK155670, and others.
Panther Classification:
Gene Ontology Categories:
