siRNA Details
| Gene Symbol: ZIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168378.1 | NP_001161850.1 | 4 | 1103 | ||
| NM_001168379.1 | NP_001161851.1 | 4 | 930 | |||
| NM_001243256.1 | NP_001230185.1 | 3 | 611 | |||
| NM_032153.5 | NP_115529.2 | 4 | 1229 | |||
| NR_033118.2 | 2 | 540 | ||||
| NR_033119.2 | 2 | 486 | ||||
| NR_040762.1 | 2 | 387 | ||||
| GenBank mRNA | AK056262.1 | - | 4 | 1040 | ||
| AK056394.1 | - | 2 | 1414 | |||
| AK094286.1 | - | 2 | 726 | |||
| AK126595.1 | - | 1 | 1766 | |||
| AK127733.1 | - | 2 | 502 | |||
| AK293981.1 | - | 4 | 930 | |||
| AK294831.1 | - | 4 | 1103 | |||
| BC126267.1 | - | 4 | 865 | 8991983 | ||
| BC136339.1 | - | 4 | 865 | 9020326 | ||
| DA106797.1 | - | 2 | 472 | |||
| DA121447.1 | - | 2 | 387 | |||
| DA218617.1 | - | 3 | 329 | |||
| DA314072.1 | - | 2 | 453 | |||
| DA411998.1 | - | 2 | 318 | |||
| DA416382.1 | - | 2 | 303 | |||

ZIC4
Genomic Map