siRNA Details
| Gene Symbol: ZNF397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135178.2 | NP_001128650.1 | 2 | 279 | ||
| NM_032347.2 | NP_115723.1 | 2 | 279 | |||
| XM_006722558.2 | XP_006722621.1 | 2 | 275 | |||
| XM_011526229.2 | XP_011524531.1 | 3 | 364 | |||
| XM_011526230.2 | XP_011524532.1 | 2 | 566 | |||
| XM_011526231.2 | XP_011524533.1 | 2 | 220 | |||
| XM_017026042.1 | XP_016881531.1 | 2 | 275 | |||
| GenBank mRNA | AF533250.2 | - | 2 | 272 | ||
| AK027384.1 | - | 2 | 254 | |||
| AY157641.2 | - | 2 | 272 | |||
| BC006172.1 | - | 2 | 254 | 4053312 | ||
| BI772202.1 | - | 2 | 278 | |||
| DB047444.1 | - | 2 | 275 | |||

ZNF397
Genomic Map