siRNA Details
| Gene Symbol: ZIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033132.3 | NP_149123.2 | 1 | 1242 | ||
| GenBank mRNA | AF378304.1 | - | 1 | 1227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZIC5| Gene Symbol: ZIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033132.3 | NP_149123.2 | 1 | 1242 | ||
| GenBank mRNA | AF378304.1 | - | 1 | 1227 | ||