siRNA Details
| Gene Symbol: ZNF101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 543 | ||
| NM_033204.3 | NP_149981.2 | 4 | 671 | |||
| GenBank mRNA | AK097169.1 | - | 4 | 663 | ||
| AL096747.1 | - | 3 | 2216 | |||
| AY149174.1 | - | 4 | 605 | |||
| BC011502.1 | - | 1 | 307 | |||
| BC094808.1 | - | 4 | 604 | |||
| BC119680.1 | - | 5 | 707 | 40116294 | ||
| CR999939.1 | - | 4 | 672 | |||
| DB264378.1 | - | 4 | 544 | |||

ZNF101
Genomic Map