siRNA Details
| Gene Symbol: TRIM47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033452.2 | NP_258411.2 | 3 | 1014 | ||
| XM_005257787.4 | XP_005257844.1 | 3 | 1061 | |||
| XM_005257788.4 | XP_005257845.1 | 3 | 420 | |||
| GenBank mRNA | AY026763.1 | - | 3 | 1015 | ||
| BC006153.1 | - | 2 | 283 | |||
| BC017299.2 | - | 3 | 446 | 5017710 | ||
| BG385464.1 | - | 3 | 579 | |||

TRIM47
Genomic Map