siRNA Details
| Gene Symbol: ZFP82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321917.1 | NP_001308846.1 | 5 | 1055 | ||
| NM_133466.3 | NP_597723.1 | 5 | 1052 | |||
| GenBank mRNA | AB075828.1 | - | 1 | 2519 | ||
| AK074942.1 | - | 5 | 1034 | |||
| AK315709.1 | - | 5 | 1012 | |||
| AL834267.1 | - | 5 | 1026 | |||
| BC032577.2 | - | 5 | 1019 | 5492906 | ||
| GU388429.1 | - | 5 | 1022 | |||

ZFP82
Genomic Map