siRNA Details
| Gene Symbol: ZNF765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001610.1 | - | 6 | 1269 | ||
| BC017357.1 | - | 6 | 1270 | |||
| Gene Symbol: ZNF331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026936.1 | XP_016882425.1 | 3 | 869 | ||

ZNF765
Genomic Map