siRNA Details
| Gene Symbol: GLI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138465.3 | NP_612474.1 | 4 | 1318 | ||
| GenBank mRNA | AY129020.1 | - | 1 | 2309 | ||
| BC014165.2 | - | 3 | 1284 | 4549240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLI4| Gene Symbol: GLI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138465.3 | NP_612474.1 | 4 | 1318 | ||
| GenBank mRNA | AY129020.1 | - | 1 | 2309 | ||
| BC014165.2 | - | 3 | 1284 | 4549240 | ||