siRNA Details
| Gene Symbol: ZNF653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138783.3 | NP_620138.2 | 6, 7 | 1603 | ||
| GenBank mRNA | AK314986.1 | - | 5, 6 | 1634 | ||
| AY072704.1 | - | 6, 7 | 1472 | |||
| BC014187.2 | - | 6, 7 | 1499 | 4634585 | ||
| BC016816.1 | - | 6, 7 | 1523 | 4094670 | ||

ZNF653
Genomic Map