siRNA Details
| Gene Symbol: ZNF625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145233.3 | NP_660276.2 | 4 | 1321 | ||
| NR_037801.1 | 4 | 1317 | ||||
| GenBank mRNA | BC007868.2 | - | 4 | 1300 | 4303049 | |
| Gene Symbol: ZNF625-ZNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK292455.1 | - | 4 | 1317 | ||

ZNF625
Genomic Map