siRNA Details
| Gene Symbol: OSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145260.2 | NP_660303.1 | 2 | 431 | ||
| XM_006711942.3 | XP_006712005.1 | 2 | 1300 | |||
| GenBank mRNA | AB082568.1 | - | 1 | 132 | ||
| AK074591.1 | - | 2 | 443 | |||
| AK122756.1 | - | 1 | 3259 | |||
| BC025712.1 | - | 2 | 431 | 5227308 | ||

OSR1
Genomic Map