siRNA Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297760.1 | NP_001284689.1 | 2, 3 | 856 | ||
| NM_018208.3 | NP_060678.2 | 2, 3 | 856 | |||
| GenBank mRNA | AB073608.1 | - | 2, 3 | 456 | ||
| AK001623.1 | - | 2, 3 | 568 | |||
| BC010082.2 | - | 2, 3 | 677 | 4329734 | ||

ETNK2
Genomic Map