siRNA Details
| Gene Symbol: ZNF684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152373.3 | NP_689586.3 | 5 | 1178 | ||
| XM_011540671.2 | XP_011538973.1 | 6 | 1511 | |||
| XM_011540672.2 | XP_011538974.1 | 6 | 1466 | |||
| XM_011540673.2 | XP_011538975.1 | 5 | 1122 | |||
| XM_017000290.1 | XP_016855779.1 | 6 | 2173 | |||
| XM_017000291.1 | XP_016855780.1 | 4 | 1075 | |||
| XM_017000292.1 | XP_016855781.1 | 4 | 1263 | |||
| GenBank mRNA | AK094072.1 | - | 5 | 1178 | ||
| BC091518.1 | - | 5 | 1082 | 30367496 | ||
| BC111463.1 | - | 1 | 517 | |||

ZNF684
Genomic Map