siRNA Details
| Gene Symbol: TMEM237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044385.2 | NP_001037850.1 | 13 | 1599 | ||
| NM_152388.3 | NP_689601.2 | 13 | 1764 | |||
| GenBank mRNA | AB053301.1 | - | 13 | 1766 | ||
| AK090601.1 | - | 13 | 1457 | |||
| AK125841.1 | - | 2 | 609 | |||
| AK304395.1 | - | 11 | 1288 | |||
| BC013730.1 | - | 5 | 682 | |||
| BC029611.1 | - | 13 | 1512 | 5304046 | ||
| BX538000.1 | - | 5 | 659 | |||

TMEM237
Genomic Map