siRNA Details
| Gene Symbol: ZNF786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152411.3 | NP_689624.2 | 2 | 264 | ||
| GenBank mRNA | AK297472.1 | - | 3 | 528 | ||
| AL834510.1 | - | 2 | 182 | |||
| BC128392.1 | - | 2 | 169 | 40007687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF786| Gene Symbol: ZNF786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152411.3 | NP_689624.2 | 2 | 264 | ||
| GenBank mRNA | AK297472.1 | - | 3 | 528 | ||
| AL834510.1 | - | 2 | 182 | |||
| BC128392.1 | - | 2 | 169 | 40007687 | ||