siRNA Details
| Gene Symbol: ZNF565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042474.2 | NP_001035939.1 | 5 | 2066 | ||
| NM_152477.4 | NP_689690.3 | 5 | 1921 | |||
| GenBank mRNA | BC068453.1 | - | 5 | 1904 | 30343899 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF565| Gene Symbol: ZNF565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042474.2 | NP_001035939.1 | 5 | 2066 | ||
| NM_152477.4 | NP_689690.3 | 5 | 1921 | |||
| GenBank mRNA | BC068453.1 | - | 5 | 1904 | 30343899 | |