siRNA Details
| Gene Symbol: ZNF48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001214906.1 | NP_001201835.1 | 3 | 1517 | ||
| NM_001214907.1 | NP_001201836.1 | 2 | 1421 | |||
| NM_001214909.1 | NP_001201838.1 | 3 | 1885 | |||
| NM_001324494.1 | NP_001311423.1 | 2 | 1791 | |||
| NM_152652.2 | NP_689865.2 | 2 | 1779 | |||
| GenBank mRNA | AK056313.1 | - | 2 | 1462 | ||
| AK122598.1 | - | 1 | 546 | |||
| AL832433.1 | - | 1 | 462 | |||
| BC007393.2 | - | 1 | 48 | |||
| BC041388.1 | - | 2 | 1466 | 5276303 | ||

ZNF48
Genomic Map