siRNA Details
| Gene Symbol: ZNF75A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302109.1 | NP_001289038.1 | 6 | 1206 | ||
| NM_001324041.1 | NP_001310970.1 | 6 | 1163 | |||
| NM_153028.3 | NP_694573.1 | 5 | 682 | |||
| NR_126069.1 | 4 | 841 | ||||
| NR_136681.1 | 5 | 1010 | ||||
| NR_136683.1 | 4 | 486 | ||||
| NR_136686.1 | 5 | 1036 | ||||
| XM_011522647.2 | XP_011520949.1 | 5 | 780 | |||
| XM_017023642.1 | XP_016879131.1 | 5 | 578 | |||
| XM_017023643.1 | XP_016879132.1 | 6 | 1567 | |||
| GenBank mRNA | AK056091.1 | - | 5 | 605 | ||
| AK093979.1 | - | 4 | 794 | |||
| BC119652.1 | - | 4 | 407 | 40115880 | ||
| BC119653.1 | - | 4 | 407 | 40115881 | ||

ZNF75A
Genomic Map