siRNA Details
| Gene Symbol: ZNF461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322821.1 | NP_001309750.1 | 5 | 555 | ||
| NM_001322823.1 | NP_001309752.1 | 5 | 497 | |||
| NM_001322825.1 | NP_001309754.1 | 5 | 518 | |||
| NM_153257.4 | NP_694989.2 | 5 | 534 | |||
| XM_017027470.1 | XP_016882959.1 | 2 | 772 | |||
| GenBank mRNA | AK292834.1 | - | 5 | 509 | ||
| AY329493.1 | - | 5 | 518 | |||
| AY500360.1 | - | 1 | 22 | |||
| BX649031.1 | - | 5 | 464 | |||
| DC328991.1 | - | 5 | 534 | |||

ZNF461
Genomic Map