siRNA Details
| Gene Symbol: ZNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301062.1 | NP_001287991.1 | 4 | 416 | ||
| NM_153608.2 | NP_705836.1 | 6 | 751 | |||
| XM_011526574.2 | XP_011524876.1 | 6 | 665 | |||
| XM_017026414.1 | XP_016881903.1 | 6 | 637 | |||
| XM_017026415.1 | XP_016881904.1 | 5 | 603 | |||
| GenBank mRNA | AK075062.1 | - | 5 | 478 | ||
| AK291575.1 | - | 6 | 821 | |||
| BC014935.2 | - | 6 | 760 | |||
| BC125068.1 | - | 1 | 477 | 40117544 | ||
| BC125069.1 | - | 4 | 346 | 40117550 | ||
| DC343694.1 | - | 5 | 516 | |||

ZNF114
Genomic Map