siRNA Details
| Gene Symbol: ZNF792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175872.4 | NP_787068.3 | 4 | 3192 | ||
| GenBank mRNA | AK095770.1 | - | 1 | 1584 | ||
| W18184.1 | - | 1 | 622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF792| Gene Symbol: ZNF792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175872.4 | NP_787068.3 | 4 | 3192 | ||
| GenBank mRNA | AK095770.1 | - | 1 | 1584 | ||
| W18184.1 | - | 1 | 622 | |||