siRNA Details
| Gene Symbol: ZNF454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178089.2 | NP_001171560.1 | 5 | 1136 | ||
| NM_001178090.2 | NP_001171561.1 | 5 | 1075 | |||
| NM_001323306.1 | NP_001310235.1 | 5 | 1133 | |||
| NM_182594.3 | NP_872400.2 | 5 | 1078 | |||
| GenBank mRNA | AK094763.1 | - | 5 | 1046 | ||
| AK292317.1 | - | 5 | 1104 | |||
| BC105061.1 | - | 4 | 838 | 8144064 | ||
| BC112276.1 | - | 4 | 840 | 8327744 | ||

ZNF454
Genomic Map