siRNA Details
| Gene Symbol: ZNF454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178089.2 | NP_001171560.1 | 5 | 2028 | ||
| NM_001178090.2 | NP_001171561.1 | 5 | 1967 | |||
| NM_001323306.1 | NP_001310235.1 | 5 | 2025 | |||
| NM_182594.3 | NP_872400.2 | 5 | 1970 | |||
| GenBank mRNA | AK094763.1 | - | 5 | 1938 | ||
| AK292317.1 | - | 5 | 1996 | |||

ZNF454
Genomic Map