siRNA Details
| Gene Symbol: NAIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197956.3 | NP_931045.1 | 2 | 1315 | ||
| GenBank mRNA | AK122729.1 | - | 2 | 1315 | ||
| AL833392.1 | - | 2 | 1107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAIF1| Gene Symbol: NAIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197956.3 | NP_931045.1 | 2 | 1315 | ||
| GenBank mRNA | AK122729.1 | - | 2 | 1315 | ||
| AL833392.1 | - | 2 | 1107 | |||