siRNA Details
| Gene Symbol: ZNF497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 2604 | ||
| NM_198458.2 | NP_940860.2 | 3 | 2701 | |||
| GenBank mRNA | AK126727.1 | - | 3 | 2607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF497| Gene Symbol: ZNF497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 2604 | ||
| NM_198458.2 | NP_940860.2 | 3 | 2701 | |||
| GenBank mRNA | AK126727.1 | - | 3 | 2607 | ||