siRNA Details
| Gene Symbol: CFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145252.1 | NP_001138724.1 | 8 | 1272 | ||
| NM_002621.2 | NP_002612.1 | 9 | 1388 | |||
| XM_017029575.1 | XP_016885064.1 | 7 | 1894 | |||
| GenBank mRNA | AK122955.1 | - | 7 | 2444 | ||
| AK310695.1 | - | 8 | 1267 | |||
| BC015756.1 | - | 8 | 1206 | 4850121 | ||
| KU178249.1 | - | 8 | 1146 | |||
| KU178250.1 | - | 9 | 1076 | |||
| M83652.1 | - | 8 | 1218 | |||
| X57748.1 | - | 8 | 1211 | |||

CFP
Genomic Map