siRNA Details
| Gene Symbol: PFDN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002624.3 | NP_002615.2 | 5 | 416 | ||
| NM_145897.2 | NP_665904.1 | 3 | 281 | |||
| GenBank mRNA | AB055803.1 | - | 5 | 325 | ||
| AB055804.1 | - | 4 | 293 | |||
| AB055805.1 | - | 3 | 190 | |||
| AK024094.1 | - | 4 | 1716 | |||
| AK292623.1 | - | 5 | 325 | |||
| BC003373.1 | - | 5 | 334 | |||
| BC062671.1 | - | 5 | 328 | 6458093 | ||
| BT007195.1 | - | 5 | 299 | |||

PFDN5
Genomic Map