siRNA Details
| Gene Symbol: SERPINA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001756.3 | NP_001747.2 | 3 | 825 | ||
| GenBank mRNA | AK225591.1 | - | 4 | 735 | ||
| AK290821.1 | - | 3 | 810 | |||
| AK313760.1 | - | 3 | 810 | |||
| BC017462.1 | - | 3 | 779 | |||
| BC056259.1 | - | 3 | 800 | 5810260 | ||
| BC058021.1 | - | 3 | 809 | 6714173 | ||
| J02943.1 | - | 2 | 756 | |||

SERPINA6
Genomic Map