siRNA Details
| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001863.4 | NP_001854.1 | 3 | 334 | ||
| GenBank mRNA | AK312140.1 | - | 3 | 231 | ||
| BC001015.2 | - | 3 | 304 | 3344701 | ||
| BC002478.2 | - | 3 | 203 | 3347455 | ||
| BT006945.1 | - | 2 | 142 | |||
| CR456789.1 | - | 2 | 142 | |||
| CR542137.1 | - | 2 | 142 | |||
| DB443544.1 | - | 3 | 334 | |||
| X13923.1 | - | 3 | 186 | |||
| X54473.1 | - | 3 | 189 | |||

COX6B1
Genomic Map