siRNA Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001867.2 | NP_001858.1 | 2 | 251 | ||
| GenBank mRNA | AK026505.1 | - | 2 | 277 | ||
| BC001005.2 | - | 2 | 251 | 2821167 | ||
| BC007498.2 | - | 2 | 215 | 3030363 | ||
| BT007098.1 | - | 2 | 162 | |||
| X16560.1 | - | 2 | 180 | |||

COX7C
Genomic Map