siRNA Details
| Gene Symbol: MYL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002841.1 | NP_001002841.1 | 8 | 775 | ||
| NM_002476.2 | NP_002467.1 | 7 | 722 | |||
| XM_005257391.4 | XP_005257448.1 | 8 | 702 | |||
| XM_011524839.2 | XP_011523141.2 | 8 | 862 | |||
| XM_017024683.1 | XP_016880172.1 | 8 | 849 | |||
| GenBank mRNA | AF116676.2 | - | 1074 | |||
| BC030228.1 | - | 7 | 711 | 5226518 | ||
| M20641.1 | - | 4 | 338 | |||
| M36172.1 | - | 7 | 703 | |||
| X13955.1 | - | 7 | 703 | |||
| X52005.1 | - | 728 | ||||

MYL4
Genomic Map