siRNA Details
| Gene Symbol: MFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166343.1 | NP_001159815.1 | 1 | 368 | ||
| NM_002405.3 | NP_002396.2 | 1 | 421 | |||
| NR_029413.1 | 1 | 421 | ||||
| GenBank mRNA | AK292123.1 | - | 1 | 362 | ||
| AK297149.1 | - | 1 | 362 | |||
| AK297833.1 | - | 1 | 354 | |||
| AK316555.1 | - | 1 | 362 | |||
| BC094814.1 | - | 1 | 382 | 5930073 | ||
| BM457814.1 | - | 1 | 369 | |||
| CR456518.1 | - | 1 | 214 | |||
| DA815501.1 | - | 1 | 421 | |||
| U94352.1 | - | 1 | 328 | |||

MFNG
Genomic Map