siRNA Details
| Gene Symbol: SLC18A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135691.2 | NP_001129163.1 | 17 | 2780 | ||
| NM_001142324.1 | NP_001135796.1 | 16 | 2684 | |||
| NM_001142325.1 | NP_001135797.1 | 16 | 2646 | |||
| NM_003053.3 | NP_003044.1 | 16 | 2576 | |||
| XM_011544625.1 | XP_011542927.1 | 15 | 2425 | |||
| XM_011544626.1 | XP_011542928.1 | 15 | 2413 | |||
| XM_011544627.1 | XP_011542929.1 | 15 | 2375 | |||
| GenBank mRNA | BC006317.2 | - | 16 | 2594 | 4125784 | |
| BC009387.2 | - | 17 | 2599 | 4127678 | ||
| U39905.1 | - | 16 | 2578 | |||

SLC18A1
Genomic Map