siRNA Details
| Gene Symbol: TM7SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277233.1 | NP_001264162.1 | 7 | 910 | ||
| NM_003273.3 | NP_003264.2 | 7 | 910 | |||
| NR_102367.1 | 6 | 855 | ||||
| GenBank mRNA | AF023676.1 | - | 5 | 1404 | ||
| AF096304.1 | - | 7 | 874 | |||
| AK290935.1 | - | 7 | 888 | |||
| AK296441.1 | - | 6 | 831 | |||
| BC009052.1 | - | 7 | 876 | 3874367 | ||
| BC012857.1 | - | 7 | 838 | 4214662 | ||
| BC038353.1 | - | 7 | 880 | 3861907 | ||

TM7SF2
Genomic Map