siRNA Details
| Gene Symbol: SLC13A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145975.1 | NP_001139447.1 | 4, 5 | 788 | ||
| NM_003984.3 | NP_003975.1 | 4, 5 | 641 | |||
| NR_027384.1 | 5, 6 | 782 | ||||
| XM_006722165.2 | XP_006722228.1 | 5, 6 | 690 | |||
| XM_011525450.2 | XP_011523752.1 | 4, 5 | 1054 | |||
| XM_011525451.1 | XP_011523753.1 | 5, 6 | 850 | |||
| XM_011525452.1 | XP_011523754.1 | 4, 5 | 641 | |||
| XM_011525453.2 | XP_011523755.1 | 4, 5 | 1062 | |||
| XM_011525454.2 | XP_011523756.1 | 4, 5 | 1076 | |||
| GenBank mRNA | AK091926.1 | - | 6, 7 | 752 | ||
| AK298388.1 | - | 4, 5 | 696 | |||
| AK303909.1 | - | 5, 6 | 690 | |||
| AK303936.1 | - | 3, 4 | 420 | |||
| AK314684.1 | - | 4, 5 | 641 | |||
| BC096276.1 | - | 5, 6 | 736 | 40006564 | ||
| BC096277.1 | - | 4, 5 | 595 | 40006565 | ||
| BC096278.1 | - | 4, 5 | 595 | 40006566 | ||
| BC096279.1 | - | 4, 5 | 595 | 40006572 | ||
| DB204108.1 | - | 3, 4 | 420 | |||
| U26209.1 | - | 4, 5 | 642 | |||

SLC13A2
Genomic Map