siRNA Details
| Gene Symbol: SLC28A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004212.3 | NP_004203.2 | 18 | 2143 | ||
| XM_011522198.2 | XP_011520500.1 | 18 | 2163 | |||
| XM_011522199.1 | XP_011520501.1 | 17 | 2015 | |||
| XM_011522202.2 | XP_011520504.1 | 10 | 1314 | |||
| GenBank mRNA | AK025121.1 | - | 2 | 1230 | ||
| AK222709.1 | - | 18 | 2143 | |||
| AK291974.1 | - | 18 | 2114 | |||
| U84392.1 | - | 18 | 2137 | |||

SLC28A2
Genomic Map