siRNA Details
| Gene Symbol: FADS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004265.3 | NP_004256.1 | 1 | 506 | ||
| GenBank mRNA | AF084559.1 | - | 1 | 298 | ||
| AF126799.1 | - | 1 | 161 | |||
| AK074939.1 | - | 1 | 238 | |||
| BC009011.1 | - | 1 | 240 | 4181037 | ||
| DA233832.1 | - | 1 | 505 | |||

FADS2
Genomic Map