siRNA Details
| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005094.3 | NP_005085.2 | 2 | 436 | ||
| XM_017014222.1 | XP_016869711.1 | 3 | 309 | |||
| GenBank mRNA | AF055899.1 | - | 1 | 151 | ||
| AK290222.1 | - | 2 | 436 | |||
| AK314681.1 | - | 2 | 276 | |||
| BC065003.1 | - | 2 | 435 | 6023438 | ||

SLC27A4
Genomic Map